Learn More
Biomatik Corporation Angiopoietin 1 (ANGPT1) anti-Human, Mouse, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510135
Encompass_Preferred
The Angiopoietin 1 (ANGPT1) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human, Mouse. Applications: WB. Alternative Names: ANG1; AGP1; AGPT; ANG-I. UniProt: Q15389. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0135 Supplier Biomatik Corporation Supplier No. CAU29138
The Angiopoietin 1 (ANGPT1) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human, Mouse. Applications: WB. Alternative Names: ANG1; AGP1; AGPT; ANG-I. UniProt: Q15389. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.