Learn More
Biomatik Corporation Cadherin, Epithelial (CDHE) anti-Human, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510137
Encompass_Preferred
The Cadherin, Epithelial (CDHE) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human. Immunogen: Pro373-Pro621. Applications: WB. Alternative Names: CD324; CDH1; Arc1; ECAD; CDH-E; E-CAD; LCAM; UVO; Cadherin 1 Type 1; Uvomorulin; E-Cadherin; Calcium-Dependent Adhesion Protein, Epithelial; Liver Cell Adhesion Molecule. UniProt: P12830. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0137 Supplier Biomatik Corporation Supplier No. CAU29141
The Cadherin, Epithelial (CDHE) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human. Immunogen: Pro373-Pro621. Applications: WB. Alternative Names: CD324; CDH1; Arc1; ECAD; CDH-E; E-CAD; LCAM; UVO; Cadherin 1 Type 1; Uvomorulin; E-Cadherin; Calcium-Dependent Adhesion Protein, Epithelial; Liver Cell Adhesion Molecule. UniProt: P12830. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.