Learn More
Biomatik Corporation Interleukin 4 (IL4) anti-Pig, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29167
Encompass_Preferred
Interleukin 4 (Il4) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Interleukin 4 (IL4). The Interleukin 4 (Il4) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: His25~Ser153. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 4 (IL4). Accession Number: P05112. Gene ID: 3565. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0142