Learn More
Biomatik Corporation Galactosidase Alpha (GLa) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29211
Encompass_Preferred
The Galactosidase Alpha (GLa) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human. Immunogen: GLa (Trp81-Leu429). Applications: WB. Alternative Names: GALA; GL-A; Alpha-D-Galactoside Galactohydrolase; Alpha-D-Galactosidase A; Melibiase; Agalsidase. UniProt: P06280. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0154