Learn More
Biomatik Corporation Interferon Beta (IFNb) anti-Chicken, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510157
Encompass_Preferred
Interferon Beta (Ifnb) Monoclonal Antibody is a Chicken (Gallus) Monoclonal Antibody to Interferon Beta (IFNb). The Interferon Beta (Ifnb) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Chicken (Gallus). Immunogen: Ser27~Gln203. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Beta (IFNb). Accession Number: Q90873. Gene ID: 554219. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0157 Supplier Biomatik Corporation Supplier No. CAU29216
Interferon Beta (Ifnb) Monoclonal Antibody is a Chicken (Gallus) Monoclonal Antibody to Interferon Beta (IFNb). The Interferon Beta (Ifnb) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Chicken (Gallus). Immunogen: Ser27~Gln203. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Beta (IFNb). Accession Number: Q90873. Gene ID: 554219. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.