Learn More
Biomatik Corporation Myeloperoxidase (MPO) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29277
Encompass_Preferred
Myeloperoxidase (Mpo) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Myeloperoxidase (MPO). The Myeloperoxidase (Mpo) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Val165~Leu258. Applications: WB, IHC, ICC, IP. Target Synonyms: Myeloperoxidase (MPO). Accession Number: P05164. Gene ID: 4353. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0166