Learn More
Biomatik Corporation Cyclin Dependent Kinase 5 (CDK5) anti-Human, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510168
Encompass_Preferred
The Cyclin Dependent Kinase 5 (CDK5) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human. Immunogen: Asp92-Gln273. Applications: WB. Alternative Names: PSSALRE; CDKN5; Cell division protein kinase 5; Serine/threonine-protein kinase PSSALRE; Tau protein kinase II catalytic subunit;TPKII catalytic subunit. UniProt: Q00535. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0168 Supplier Biomatik Corporation Supplier No. CAU29292
The Cyclin Dependent Kinase 5 (CDK5) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human. Immunogen: Asp92-Gln273. Applications: WB. Alternative Names: PSSALRE; CDKN5; Cell division protein kinase 5; Serine/threonine-protein kinase PSSALRE; Tau protein kinase II catalytic subunit;TPKII catalytic subunit. UniProt: Q00535. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.