Learn More
Biomatik Corporation Gonadotropin Releasing Hormone (GnRH) anti-Rat, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510172
Encompass_Preferred
The Gonadotropin Releasing Hormone (GnRH) Monoclonal Antibody is raised in Mouse. Species Reactivity: Rat. Immunogen: His25-Met92. Applications: WB. Alternative Names: GNRH1; GRH; LNRH; LHRH; LH-RH I; Luliberin; Luteinizing-Hormone Releasing Hormone; Progonadoliberin-1; Gonadoliberin I; Gonadorelin; GnRH-associated peptide 1. UniProt: P07490. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0172 Supplier Biomatik Corporation Supplier No. CAU29307
The Gonadotropin Releasing Hormone (GnRH) Monoclonal Antibody is raised in Mouse. Species Reactivity: Rat. Immunogen: His25-Met92. Applications: WB. Alternative Names: GNRH1; GRH; LNRH; LHRH; LH-RH I; Luliberin; Luteinizing-Hormone Releasing Hormone; Progonadoliberin-1; Gonadoliberin I; Gonadorelin; GnRH-associated peptide 1. UniProt: P07490. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.