Learn More
Biomatik Corporation Procollagen I N-Terminal Propeptide (PINP) anti-Human, Rat, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510177
Encompass_Preferred
Procollagen I N-Terminal Propeptide (Pinp) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Procollagen I N-Terminal Propeptide (PINP). The Procollagen I N-Terminal Propeptide (Pinp) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Gln23~Pro161. Applications: WB, IHC. Target Synonyms: Procollagen I N-Terminal Propeptide (PINP). Accession Number: P02452. Gene ID: 1277. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0177 Supplier Biomatik Corporation Supplier No. CAU29330
Procollagen I N-Terminal Propeptide (Pinp) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Procollagen I N-Terminal Propeptide (PINP). The Procollagen I N-Terminal Propeptide (Pinp) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Gln23~Pro161. Applications: WB, IHC. Target Synonyms: Procollagen I N-Terminal Propeptide (PINP). Accession Number: P02452. Gene ID: 1277. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.