Learn More
Biomatik Corporation Interleukin 33 (IL33) anti-Human, Rat, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29393
Encompass_Preferred
Interleukin 33 (Il33) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Interleukin 33 (IL33). The Interleukin 33 (Il33) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ser112~Thr228. Applications: WB, IHC. Target Synonyms: Interleukin 33 (IL33). Accession Number: O95760. Gene ID: 90865. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0189