Learn More
Biomatik Corporation Toll Like Receptor 5 (TLR5) anti-Human, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510190
Encompass_Preferred
Toll Like Receptor 5 (Tlr5) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Toll Like Receptor 5 (TLR5). The Toll Like Receptor 5 (Tlr5) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Tyr693~Ser858. Applications: WB, IHC, ICC, IP. Target Synonyms: Toll Like Receptor 5 (TLR5). Accession Number: O60602. Gene ID: 7100. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0190 Supplier Biomatik Corporation Supplier No. CAU29396
Toll Like Receptor 5 (Tlr5) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Toll Like Receptor 5 (TLR5). The Toll Like Receptor 5 (Tlr5) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Tyr693~Ser858. Applications: WB, IHC, ICC, IP. Target Synonyms: Toll Like Receptor 5 (TLR5). Accession Number: O60602. Gene ID: 7100. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.