Learn More
Biomatik Corporation Mucin 5 Subtype AC (MUC5AC) anti-Human, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510200
Encompass_Preferred
Mucin 5 Subtype Ac (Muc5Ac) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Mucin 5 Subtype AC (MUC5AC). The Mucin 5 Subtype Ac (Muc5Ac) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Tyr5568~His5654. Applications: WB. Target Synonyms: Mucin 5 Subtype AC (MUC5AC). Accession Number: P98088. Gene ID: 4586. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0200 Supplier Biomatik Corporation Supplier No. CAU29485
Mucin 5 Subtype Ac (Muc5Ac) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Mucin 5 Subtype AC (MUC5AC). The Mucin 5 Subtype Ac (Muc5Ac) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Tyr5568~His5654. Applications: WB. Target Synonyms: Mucin 5 Subtype AC (MUC5AC). Accession Number: P98088. Gene ID: 4586. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.