Learn More
Biomatik Corporation Interleukin 10 (IL10) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29519
Encompass_Preferred
Interleukin 10 (Il10) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Interleukin 10 (IL10). The Interleukin 10 (Il10) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met1~Asn178. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 10 (IL10). Accession Number: P22301. Gene ID: 3586. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0205