Learn More
Biomatik Corporation Receptor Activator of Nuclear Factor Kappa B (RANk) anti-Human, Rat, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510211
Encompass_Preferred
Receptor Activator Of Nuclear Factor Kappa B (Rank) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Receptor Activator Of Nuclear Factor Kappa B (RANk). The Receptor Activator Of Nuclear Factor Kappa B (Rank) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Val330~Lys615. Applications: WB. Target Synonyms: Receptor Activator Of Nuclear Factor Kappa B (RANk). Accession Number: Q9Y6Q6. Gene ID: 8792. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0211 Supplier Biomatik Corporation Supplier No. CAU29557
Receptor Activator Of Nuclear Factor Kappa B (Rank) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Receptor Activator Of Nuclear Factor Kappa B (RANk). The Receptor Activator Of Nuclear Factor Kappa B (Rank) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Val330~Lys615. Applications: WB. Target Synonyms: Receptor Activator Of Nuclear Factor Kappa B (RANk). Accession Number: Q9Y6Q6. Gene ID: 8792. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.