Learn More
Biomatik Corporation Ki-67 Protein (Ki67P) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29568
Encompass_Preferred
Ki-67 Protein (Ki-67) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Ki-67 Protein (Ki-67). The Ki-67 Protein (Ki-67) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Gly3088~Lys3235. Applications: WB, IHC, ICC, IP. Target Synonyms: Ki-67 Protein (Ki-67). Accession Number: P46013. Gene ID: 4288. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0212