Learn More
Biomatik Corporation Lecithin Cholesterol Acyltransferase (LCAT) anti-Human, Mouse Monoclonal Antibody (100UG)
Catalog No. 501510217
Encompass_Preferred
Lecithin Cholesterol Acyltransferase (Lcat) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Lecithin Cholesterol Acyltransferase (LCAT). The Lecithin Cholesterol Acyltransferase (Lcat) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ile290~Ser433. Applications: WB, IHC. Target Synonyms: Lecithin Cholesterol Acyltransferase (LCAT). Accession Number: P04180. Gene ID: 3931. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0217 Supplier Biomatik Corporation Supplier No. CAU29674
Lecithin Cholesterol Acyltransferase (Lcat) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Lecithin Cholesterol Acyltransferase (LCAT). The Lecithin Cholesterol Acyltransferase (Lcat) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ile290~Ser433. Applications: WB, IHC. Target Synonyms: Lecithin Cholesterol Acyltransferase (LCAT). Accession Number: P04180. Gene ID: 3931. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.