Learn More
Biomatik Corporation Thymidine Kinase 1, Soluble (TK1) anti-Human, Pig, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29678
Encompass_Preferred
Thymidine Kinase 1, Soluble (Tk1) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Thymidine Kinase 1, Soluble (TK1). The Thymidine Kinase 1, Soluble (Tk1) Monoclonal Antibody has been validated for the following applications: WB, ICC/IF. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ser2~Asn234. Applications: WB, ICC/IF. Target Synonyms: Thymidine Kinase 1, Soluble (TK1). Accession Number: P04183. Gene ID: 7083. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0219