Learn More
Biomatik Corporation Galectin 7 (GAL7) anti-Human, Rat, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29679
Encompass_Preferred
The Galectin 7 (GAL7) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human, Rat. Immunogen: GAL7 (Met1-Phe136). Applications: WB, IHC. Alternative Names: LGALS7; PIG1; TP53I1; HKL-14; Lectin Galactoside-Binding Soluble 7; p53-induced gene 1 protein. UniProt: P47929. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0220