Learn More
Biomatik Corporation POLH anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08013
Encompass_Preferred
Polh Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to POLH. The Polh Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dna Polymerase Eta Protein (408-609Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: POLH. Accession Number: Q9Y253. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7598