Learn More
Biomatik Corporation MAN2A1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08042
Encompass_Preferred
Man2A1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MAN2A1. The Man2A1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Alpha-Mannosidase 2 Protein (31-181Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MAN2A1. Accession Number: Q16706. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7626