Learn More
Biomatik Corporation TRIP4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08046
Encompass_Preferred
Trip4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIP4. The Trip4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Activating Signal Cointegrator 1 Protein (360-535Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TRIP4. Accession Number: Q15650. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7630