Learn More
Biomatik Corporation ALX1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08050
Encompass_Preferred
The ALX1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human ALX homeobox protein 1 protein (10-304AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: ALX homeobox protein 1, Cartilage homeoprotein 1, ALX1. UniProt: Q15699. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7634