Learn More
Biomatik Corporation EIF3M anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507635
Encompass_Preferred
The EIF3M Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Eukaryotic translation initiation factor 3 subunit M protein (2-374AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Eukaryotic translation initiation factor 3 subunit M, Fetal lung protein B5, hFL-B5, PCI domain-containing protein 1, EIF3M. UniProt: Q7L2H7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7635 Supplier Biomatik Corporation Supplier No. CAC08051
The EIF3M Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Eukaryotic translation initiation factor 3 subunit M protein (2-374AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Eukaryotic translation initiation factor 3 subunit M, Fetal lung protein B5, hFL-B5, PCI domain-containing protein 1, EIF3M. UniProt: Q7L2H7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.