Learn More
Biomatik Corporation IL34 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08052
Encompass_Preferred
Il34 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL34. The Il34 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-34 Protein (191-240Aa). Applications: ELISA, WB, IHC. Target Synonyms: IL34. Accession Number: Q6ZMJ4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7636