Learn More
Biomatik Corporation RBFA anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08065
Encompass_Preferred
Rbfa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RBFA. The Rbfa Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Putative Ribosome-Binding Factor A, Mitochondrial Protein (201-343Aa). Applications: ELISA, WB, IHC. Target Synonyms: RBFA. Accession Number: Q8N0V3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7649