Learn More
Biomatik Corporation GPER1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507660
Encompass_Preferred
The GPER1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human G-protein coupled estrogen receptor 1 protein (1-62AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: G-protein coupled estrogen receptor 1, Chemoattractant receptor-like 2, Flow-induced endothelial G-protein coupled receptor 1, FEG-1, G protein-coupled estrogen receptor 1, G-protein coupled receptor 30, GPCR-Br, IL8-related receptor DRY12, Lymphocyte-derived G-protein coupled receptor, LYGPR, Membrane estrogen receptor, mER, GPER1, CEPR, CMKRL2, DRY12, GPER, GPR30. UniProt: Q99527. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7660 Supplier Biomatik Corporation Supplier No. CAC08076
The GPER1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human G-protein coupled estrogen receptor 1 protein (1-62AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: G-protein coupled estrogen receptor 1, Chemoattractant receptor-like 2, Flow-induced endothelial G-protein coupled receptor 1, FEG-1, G protein-coupled estrogen receptor 1, G-protein coupled receptor 30, GPCR-Br, IL8-related receptor DRY12, Lymphocyte-derived G-protein coupled receptor, LYGPR, Membrane estrogen receptor, mER, GPER1, CEPR, CMKRL2, DRY12, GPER, GPR30. UniProt: Q99527. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.