Learn More
Biomatik Corporation VPS29 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08087
Encompass_Preferred
Vps29 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VPS29. The Vps29 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Vacuolar Protein Sorting-Associated Protein 29 Protein (68-145Aa). Applications: ELISA, WB. Target Synonyms: VPS29. Accession Number: Q9UBQ0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7671