Learn More
Biomatik Corporation SHOC2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507672
Encompass_Preferred
Shoc2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SHOC2. The Shoc2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Leucine-Rich Repeat Protein Shoc-2 Protein (3-89Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SHOC2. Accession Number: Q9UQ13. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7672 Supplier Biomatik Corporation Supplier No. CAC08088
Shoc2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SHOC2. The Shoc2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Leucine-Rich Repeat Protein Shoc-2 Protein (3-89Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SHOC2. Accession Number: Q9UQ13. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.