Learn More
Biomatik Corporation HS6ST1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08095
Encompass_Preferred
The HS6ST1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Heparan-sulfate 6-O-sulfotransferase 1 protein (3-133AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:1000-1:5000, IF:1:50-1:200. Alternative Names: Heparan-sulfate 6-O-sulfotransferase 1, HS6ST1, HS6ST. UniProt: O60243. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7679