Learn More
Biomatik Corporation PDGFRA anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08107
Encompass_Preferred
Pdgfra Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PDGFRA. The Pdgfra Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Platelet-Derived Growth Factor Receptor Alpha Protein (30-198Aa). Applications: ELISA, WB, IF. Target Synonyms: PDGFRA. Accession Number: P16234. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7691