Learn More
Biomatik Corporation SLC35A2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507693
Encompass_Preferred
Slc35A2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC35A2. The Slc35A2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Udp-Galactose Translocator Protein (136-234Aa). Applications: ELISA, WB, IF. Target Synonyms: SLC35A2. Accession Number: P78381. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7693 Supplier Biomatik Corporation Supplier No. CAC08109
Slc35A2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC35A2. The Slc35A2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Udp-Galactose Translocator Protein (136-234Aa). Applications: ELISA, WB, IF. Target Synonyms: SLC35A2. Accession Number: P78381. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.