Learn More
Biomatik Corporation HIF1A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07003
Encompass_Preferred
The HIF1A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Hypoxia-inducible factor 1-alpha protein (579-826AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, Class E basic helix-loop-helix protein 78, Member of PAS protein 1, PAS domain-containing protein 8.HIF1A, BHLHE78, MOP1, PASD8. UniProt: Q16665. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6590