Learn More
Biomatik Corporation TNF anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07005
Encompass_Preferred
Tnf Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TNF. The Tnf Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tumor Necrosis Factor Protein (77-233Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TNF. Accession Number: P01375. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6592