Learn More
Biomatik Corporation AKT1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07008
Encompass_Preferred
The AKT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human RAC-alpha serine/threonine-protein kinase protein (1-480AA). Applications: ELISA, IHC, IF, ChIP. Recommended dilution: IHC:1:20-1:200, IF:1:100-1:500. Alternative Names: Protein kinase B Protein kinase B alpha Proto-oncogene c-Akt RAC-PK-alpha AKT1 PKB, RAC. UniProt: P31749. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6595