Learn More
Biomatik Corporation ACTB anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506598
Encompass_Preferred
The ACTB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Actin, cytoplasmic 1 protein (102-375AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Beta-actin, ACTB. UniProt: P60709. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6598 Supplier Biomatik Corporation Supplier No. CAC07011
The ACTB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Actin, cytoplasmic 1 protein (102-375AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Beta-actin, ACTB. UniProt: P60709. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.