Learn More
Biomatik Corporation CXCR4 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506602
Encompass_Preferred
The CXCR4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human C-X-C chemokine receptor type 4 protein (1-356AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: C-X-C chemokine receptor type 4, CXC-R4, FB22, Leukocyte-derived seven transmembrane domain receptor, CXCR4. UniProt: P61073. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6602 Supplier Biomatik Corporation Supplier No. CAC07015
The CXCR4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human C-X-C chemokine receptor type 4 protein (1-356AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: C-X-C chemokine receptor type 4, CXC-R4, FB22, Leukocyte-derived seven transmembrane domain receptor, CXCR4. UniProt: P61073. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.