Learn More
Biomatik Corporation CASP8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506606
Encompass_Preferred
The CASP8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Caspase-8 protein (217-374AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Apoptotic cysteine protease, Apoptotic protease Mch-5, CAP4, FADD-homologous ICE/ced-3-like protease, FADD-like ICE, CASP8, MCH5. UniProt: Q14790. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6606 Supplier Biomatik Corporation Supplier No. CAC07019
The CASP8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Caspase-8 protein (217-374AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Apoptotic cysteine protease, Apoptotic protease Mch-5, CAP4, FADD-homologous ICE/ced-3-like protease, FADD-like ICE, CASP8, MCH5. UniProt: Q14790. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.