Learn More
Biomatik Corporation HMGB1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07036
Encompass_Preferred
The HMGB1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human High mobility group protein B1 protein (8-179AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. UniProt: P09429. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6623