Learn More
Biomatik Corporation CDC42 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07038
Encompass_Preferred
The CDC42 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cell division control protein 42 homolog protein (2-188AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Cell division control protein 42 homolog, G25K GTP-binding protein, CDC42. UniProt: P60953. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6625