Learn More
Biomatik Corporation KEAP1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506626
Encompass_Preferred
Keap1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KEAP1. The Keap1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Kelch-Like Ech-Associated Protein 1 Protein (1-624Aa). Applications: ELISA, WB, IHC. Target Synonyms: KEAP1. Accession Number: Q14145. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6626 Supplier Biomatik Corporation Supplier No. CAC07039
Keap1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KEAP1. The Keap1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Kelch-Like Ech-Associated Protein 1 Protein (1-624Aa). Applications: ELISA, WB, IHC. Target Synonyms: KEAP1. Accession Number: Q14145. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.