Learn More
Biomatik Corporation FGF2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07072
Encompass_Preferred
The FGF2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Fibroblast growth factor 2 protein (143-288AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: FGFB, FGF2, Basic fibroblast growth factor, bFGF, Heparin-binding growth factor 2, HBGF-2, FGF-2. UniProt: P09038. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6659