Learn More
Biomatik Corporation TGFBR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07073
Encompass_Preferred
Tgfbr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TGFBR1. The Tgfbr1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tgf-Beta Receptor Type-1 Protein (157-244Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TGFBR1. Accession Number: P36897. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6660