Learn More
Biomatik Corporation LDHA anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506661
Encompass_Preferred
Ldha Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LDHA. The Ldha Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human L-Lactate Dehydrogenase A Chain Protein (5-323Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LDHA. Accession Number: P00338. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6661 Supplier Biomatik Corporation Supplier No. CAC07074
Ldha Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LDHA. The Ldha Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human L-Lactate Dehydrogenase A Chain Protein (5-323Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LDHA. Accession Number: P00338. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.