Learn More
Biomatik Corporation Casp12 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506664
Encompass_Preferred
The Casp12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse Caspase-12 protein (1-92AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: CASP-12, Casp12. UniProt: O08736. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6664 Supplier Biomatik Corporation Supplier No. CAC07077
The Casp12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse Caspase-12 protein (1-92AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: CASP-12, Casp12. UniProt: O08736. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.