Learn More
Biomatik Corporation CYP2E1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07078
Encompass_Preferred
The CYP2E1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cytochrome P450 2E1 protein (49-288AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Cytochrome P450 2E1, 4-nitrophenol 2-hydroxylase, CYPIIE1, Cytochrome P450-J, CYP2E1, CYP2E. UniProt: P05181. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6665