Learn More
Biomatik Corporation BMP2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07085
Encompass_Preferred
The BMP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Bone morphogenetic protein 2 protein (283-396AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Bone morphogenetic protein 2A, BMP2, BMP2A. UniProt: P12643. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6672