Learn More
Biomatik Corporation USP30 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506674
Encompass_Preferred
Usp30 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to USP30. The Usp30 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquitin Carboxyl-Terminal Hydrolase 30 Protein (101-400Aa). Applications: ELISA, IHC. Target Synonyms: USP30. Accession Number: Q70CQ3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6674 Supplier Biomatik Corporation Supplier No. CAC07087
Usp30 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to USP30. The Usp30 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquitin Carboxyl-Terminal Hydrolase 30 Protein (101-400Aa). Applications: ELISA, IHC. Target Synonyms: USP30. Accession Number: Q70CQ3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.