Learn More
Biomatik Corporation HIF1AN anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07088
Encompass_Preferred
The HIF1AN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Hypoxia-inducible factor 1-alpha inhibitor protein (2-254AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200, IP:1:200-1:2000. Alternative Names: Hypoxia-inducible factor 1-alpha inhibitor (EC:1.14.11.30, EC:1.14.11.n4), HIF1AN, FIH1, Factor inhibiting HIF-1, FIH-1, Hypoxia-inducible factor asparagine hydroxylase. UniProt: Q9NWT6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6675