Learn More
Biomatik Corporation KRT8 anti-Human, Zebrafish, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07095
Encompass_Preferred
Krt8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KRT8. The Krt8 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Keratin, Type Ii Cytoskeletal 8 Protein (2-483Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: KRT8. Accession Number: P05787. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6682