Learn More
Biomatik Corporation RHOA anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506687
Encompass_Preferred
Rhoa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RHOA. The Rhoa Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transforming Protein Rhoa Protein (1-191Aa). Applications: ELISA, IHC. Target Synonyms: RHOA. Accession Number: P61586. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6687 Supplier Biomatik Corporation Supplier No. CAC07100
Rhoa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RHOA. The Rhoa Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transforming Protein Rhoa Protein (1-191Aa). Applications: ELISA, IHC. Target Synonyms: RHOA. Accession Number: P61586. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.